Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human CD38, BD Horizon™
Mouse Monoclonal Antibody
$175.50 - $447.50
Specifications
| Antigen | CD38 |
|---|---|
| Clone | HIT2 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RB744 |
Description
CD38 Monoclonal antibody specifically detects CD38 in Human samples. It is validated for Flow cytometrySpecifications
| CD38 | |
| Flow Cytometry | |
| RB744 | |
| Mouse | |
| Human | |
| T10; ADP-ribosyl cyclase 1; Cyclic ADP-ribose hydrolase 1; gp45 | |
| IgG1 κ | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| HIT2 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| 952 | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title