Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human CD62L (L-Selectin), BD Horizon™
Mouse Monoclonal Antibody
$186.00 - $432.00
Specifications
Antigen | CD62L (L-Selectin) |
---|---|
Clone | DREG-56 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB744 |
Description
CD62L (L-Selectin) Monoclonal antibody specifically detects CD62L (L-Selectin) in Human samples. It is validated for Flow cytometrySpecifications
CD62L (L-Selectin) | |
Flow Cytometry | |
RB744 | |
Mouse | |
Human | |
SELL; L-selectin; LSEL; LAM-1; LECAM-1; LEU8; LNHR; MEL-14; PLNHR; TQ-1 | |
Supernatant from PMA-activated Human Peripheral Blood Leukocytes | |
Affinity Chromatography |
DREG-56 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
6402 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title