Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human CD8, BD Horizon™
Mouse Monoclonal Antibody
$152.00 - $376.00
Specifications
Antigen | CD8 |
---|---|
Clone | RPA-T8 |
Applications | Flow Cytometry |
Classification | Monoclonal |
Conjugate | RB744 |
Description
CD8 Monoclonal antibody specifically detects CD8 in Human samples. It is validated for Flow cytometrySpecifications
CD8 | |
Flow Cytometry | |
RB744 | |
Mouse | |
Human | |
CD8α; CD8A; CD8 alpha; Leu2; MAL; T8; p32 | |
Human CD8a | |
Affinity Chromatography |
RPA-T8 | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
925 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title