Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Human FoxP3, BD Horizon™
Mouse Monoclonal Antibody
$186.20 - $458.00
Specifications
| Antigen | FoxP3 |
|---|---|
| Clone | 259D/C7 |
| Applications | Flow Cytometry |
| Classification | Monoclonal |
| Conjugate | RB744 |
Description
FoxP3 Monoclonal antibody specifically detects FoxP3 in Human samples. It is validated for Intracellular staining (flow cytometry)Specifications
| FoxP3 | |
| Flow Cytometry | |
| RB744 | |
| Mouse | |
| Human | |
| scurfin; IPEX; JM2; AIID; XPID; DIETER; PIDX | |
| IgG1 | |
| Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
| 259D/C7 | |
| Monoclonal | |
| Liquid | |
| RUO | |
| Aqueous buffered solution containing ≤0.09% sodium azide. | |
| FoxP3 | |
| Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title