Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB744 Mouse Anti-Human TCR γδ , BD Horizon™

Catalog No. BDB570494 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570494 25 Tests
BDB570495 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570494 Supplier BD Biosciences Supplier No. 570494
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

TCR γδ Monoclonal antibody specifically detects TCR γδ in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen TCR γδ
Applications Flow Cytometry
Classification Monoclonal
Clone 11F2
Conjugate RB744
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias TCRgd; γδ TCR; TRG@/TRD@; TCRG/TCRD; TCR gamma delta
Host Species Mouse
Immunogen Sepharose™ bead/CD3/γ/δ TCR complex
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Gene ID (Entrez) 6964, 6965
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.