Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB744 Mouse Anti-Mouse CD45.2, BD OptiBuild™

Catalog No. BDB758138 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB758138 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB758138 Supplier BD Biosciences Supplier No. 758138
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD45.2 Monoclonal antibody specifically detects CD45.2 in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD45.2
Applications Flow Cytometry
Classification Monoclonal
Clone 104
Concentration 0.2 mg/mL
Conjugate RB744
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Ly-5.2; T200; LCA; Leukocyte common antigen; Ptprc
Host Species Mouse
Immunogen B10.S mouse thymocytes and splenocytes
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Gene ID (Entrez) 19264
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.