Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Mouse H-2Kq, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 756889
Description
H-2Kq Monoclonal antibody specifically detects H-2Kq in Mouse samples. It is validated for Flow cytometry
Specifications
H-2Kq | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
KH114 | |
RB744 | |
H-2Kq; H2-Kq; Histocompatibility-2Kq | |
B10.Q mouse skin graft and splenocytes | |
50 μg | |
14972 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction