Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Mouse Vβ 12 T-Cell Receptor, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757038
Description
Vβ 12 T-Cell Receptor Monoclonal antibody specifically detects Vβ 12 T-Cell Receptor in Mouse samples. It is validated for Flow cytometry
Specifications
Vβ 12 T-Cell Receptor | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
MR11-1 | |
RB744 | |
TCR Vβ12; TCR V beta 12; TCR Vb12 | |
Not Reported | |
50 μg | |
110360 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction