Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Mouse Anti-Rat CD71, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 757850
Description
CD71 Monoclonal antibody specifically detects CD71 in Rat samples. It is validated for Flow cytometry
Specifications
CD71 | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Rat | |
Liquid |
Flow Cytometry | |
OX-26 | |
RB744 | |
CD71; Transferrin Receptor; TfR; TfR1; Tfrc; Trfr; TR | |
PHA-activated PVG rat lymph node cells | |
50 μg | |
64678 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction