Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Rat Anti-Mouse CD115 (CSF-1 Receptor), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 757327
Description
CD115 (CSF-1 Receptor) Monoclonal antibody specifically detects CD115 (CSF-1 Receptor) in Mouse samples. It is validated for Flow cytometry
Specifications
CD115 (CSF-1 Receptor) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
AFS98 (also known as AFS98) | |
RB744 | |
CD115; CSF-1R; Csf1r; M-CSFR; Fms; c-fms | |
Mouse | |
50 μg | |
12978 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction