Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Rat Anti-Mouse CD19, BD Horizon™
Rat Monoclonal Antibody
$98.00 - $252.00
Specifications
Antigen | CD19 |
---|---|
Clone | 1D3 |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
CD19 Monoclonal antibody specifically detects CD19 in Mouse samples. It is validated for Flow cytometrySpecifications
CD19 | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
12478 | |
IgG2a κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
1D3 | |
Flow Cytometry | |
RB744 | |
Rat | |
Mouse | |
Cd19; CD19 antigen; B-lymphocyte antigen CD19 | |
Mouse CD19 Transfected Cell Line | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title