Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB744 Rat Anti-Mouse CD26, BD OptiBuild™

Catalog No. BDB757372 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB757372 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. BDB757372 Supplier BD Biosciences Supplier No. 757372
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD26 Monoclonal antibody specifically detects CD26 in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD26
Applications Flow Cytometry
Classification Monoclonal
Clone H194-112
Concentration 0.2 mg/mL
Conjugate RB744
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Cd26; Dpp4; dipeptidylpeptidase 4; THAM, DPP IV
Host Species Rat
Immunogen Cell-surface molecules solubilized from BALB/c mouse thymocytes
Purification Method Affinity Chromatography
Quantity 50 μg
Regulatory Status RUO
Gene ID (Entrez) 13482
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.