Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Rat Anti-Mouse CD357 (GITR), BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 756843
Description
CD357 (GITR) Monoclonal antibody specifically detects CD357 (GITR) in Mouse samples. It is validated for Flow cytometry
Specifications
CD357 (GITR) | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
DTA-1 (also known as DTA-1) | |
RB744 | |
GITR; Gitr; glucocorticoid-induced TNFR-related protein; Tnfrsf18; AITR | |
Mouse CD25+ CD4+ T Cell Line | |
50 μg | |
21936 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. | |
IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction