Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Rat Anti-Mouse Ig λ1 λ2 & λ3 Light Chain
Rat Monoclonal Antibody
Supplier: BD Biosciences 757920
Description
Ig λ1 λ2 & λ3 Light Chain Monoclonal antibody specifically detects Ig λ1 λ2 & λ3 Light Chain in Mouse samples. It is validated for Flow cytometry
Specifications
Ig λ1 λ2 & λ3 Light Chain | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2a κ |
Flow Cytometry | |
R26-46 | |
RB744 | |
Igl; Ig λ; Immunoglobulin lambda chain complex; Ig λ1/ λ2/ λ3 | |
Pooled Mouse Ig | |
50 μg | |
Mouse | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction