Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB744 Rat Anti-Mouse Vβ4 T-Cell Receptor, BD OptiBuild™
Rat Monoclonal Antibody
Supplier: BD Biosciences 757536
Description
Vβ4 T-Cell Receptor Monoclonal antibody specifically detects Vβ4 T-Cell Receptor in Mouse samples. It is validated for Flow cytometry
Specifications
Vβ4 T-Cell Receptor | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Rat | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
KT4 | |
RB744 | |
TCR V beta 4; TCR Vβ4 | |
KT4 B10.D2 mouse T-cell clone I3 | |
50 μg | |
21577 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG2b κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction