Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB780 Mouse Anti-Human CD39 (ENTPD1), BD Horizon™

Catalog No. BDB570123 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570123 25 Tests
BDB570042 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570123 Supplier BD Biosciences Supplier No. 570123
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

CD39 (ENTPD1) Monoclonal antibody specifically detects CD39 (ENTPD1) in Human samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD39 (ENTPD1)
Applications Flow Cytometry
Classification Monoclonal
Clone A1 (also known as A1/CD39)
Conjugate RB780
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias A1; ENTPD1; NTPDase-1; ecto-ATPDase 1; ecto-ATPase 1; ecto-apyrase
Host Species Mouse
Immunogen PHA-activated Human Peripheral Blood Mononuclear Cells
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.