Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB780 Mouse Anti-Insulin, BD Horizon™

Mouse Monoclonal Antibody

$130.00 - $326.00

Specifications

Antigen Insulin
Clone T56-706
Concentration 0.2 mg/mL
Applications Flow Cytometry
Classification Monoclonal
View More Specs

Products 2
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Quantity Price Quantity & Availability  
BDB570372
View Documents
BD Biosciences
570372
0.1 mg
Each for $326.00
Only null left
Add to Cart
 
BDB570442
View Documents
BD Biosciences
570442
25 μg
Each for $130.00
Only null left
Add to Cart
 
Description

Description

Insulin Monoclonal antibody specifically detects Insulin in Human samples. It is validated for Intracellular staining (flow cytometry)
Specifications

Specifications

Insulin
0.2 mg/mL
Monoclonal
Liquid
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
3630, 16334, 24506
IgG1 κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
T56-706
Flow Cytometry
RB780
Mouse
Human
INS; IDDM; ILPR; IRDN; IDDM1; IDDM2; MODY10; Ins2
Mature human insulin (A- and B-chain) Recombinant Protein
Affinity Chromatography
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Your feedback has been submitted: Thank you for helping us improve our website.