Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB780 Mouse Anti-Mouse H-2Kb/SIINFEKL, BD Horizon™
Mouse Monoclonal Antibody
$130.00 - $326.00
Specifications
Antigen | H-2Kb/SIINFEKL |
---|---|
Clone | 25-D1.16.rMAb (also known as 25-D1.16) |
Concentration | 0.2 mg/mL |
Applications | Flow Cytometry |
Classification | Monoclonal |
Description
H-2Kb/SIINFEKL Monoclonal antibody specifically detects H-2Kb/SIINFEKL in Mouse samples. It is validated for Flow cytometrySpecifications
H-2Kb/SIINFEKL | |
0.2 mg/mL | |
Monoclonal | |
Liquid | |
RUO | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
14972 | |
IgG1 κ | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. |
25-D1.16.rMAb (also known as 25-D1.16) | |
Flow Cytometry | |
RB780 | |
Mouse | |
Mouse | |
H-2Kb/SIINFEKL; H-2Kb:SIINFEKL; SIINFEKL-bound H-2Kb | |
SIINFEKL-pulsed RMA-S cells | |
Affinity Chromatography |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title