Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RB780 Mouse Anti-Mouse TCR Vδ6B, BD OptiBuild™
Mouse Monoclonal Antibody
Supplier: BD Biosciences 758033
Description
TCR Vδ6B Monoclonal antibody specifically detects TCR Vδ6B in Mouse samples. It is validated for Flow cytometry
Specifications
TCR Vδ6B | |
Monoclonal | |
0.2 mg/mL | |
Aqueous buffered solution containing ≤0.09% sodium azide. | |
Mouse | |
Affinity Chromatography | |
RUO | |
Mouse | |
Liquid |
Flow Cytometry | |
F4.22.rMAb (also known as Anti-Vδ6 λ12) | |
RB780 | |
TCR Variable delta 6B; TCR Vd6b; TCR Vδ6 λ12 | |
Mouse γδ T Cell Hybridoma Cells | |
50 μg | |
110371 | |
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated to the dye under optimum conditions that minimize unconjugated dye and antibody. | |
IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction