Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB780 Rat Anti-Human IL-6, BD Horizon™

Catalog No. BDB570148 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
100 Tests
25 Tests
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570148 25 Tests
BDB570067 100 Tests
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570148 Supplier BD Biosciences Supplier No. 570148
Add to Cart
Add to Cart

Rat Monoclonal Antibody

IL-6 Monoclonal antibody specifically detects IL-6 in Human samples. It is validated for Intracellular staining (flow cytometry)
TRUSTED_SUSTAINABILITY

Specifications

Antigen IL-6
Applications Flow Cytometry
Classification Monoclonal
Clone MQ2-13A5
Conjugate RB780
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias IL6; Interleukin-6; BSF-2; CDF; HGF; HSF; IFNB2
Host Species Rat
Immunogen Human IL-6 Recombinant Protein
Purification Method Affinity Chromatography
Quantity 25 Tests
Regulatory Status RUO
Gene ID (Entrez) 3569
Target Species Human
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.