Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RB780 Rat Anti-Mouse CD25 (IL-2 Receptor α), BD Horizon™

Catalog No. BDB570356 Shop All BD Biosciences Products
Change view
Click to view available options
Quantity:
0.1 mg
25 μg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
BDB570356 0.1 mg
BDB570426 25 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. BDB570356 Supplier BD Biosciences Supplier No. 570356
Add to Cart
Add to Cart

Rat Monoclonal Antibody

CD25 (IL-2 Receptor α) Monoclonal antibody specifically detects CD25 (IL-2 Receptor α) in Mouse samples. It is validated for Flow cytometry
TRUSTED_SUSTAINABILITY

Specifications

Antigen CD25 (IL-2 Receptor α)
Applications Flow Cytometry
Classification Monoclonal
Clone PC61
Concentration 0.2 mg/mL
Conjugate RB780
Formulation Aqueous buffered solution containing ≤0.09% sodium azide.
Gene Alias Interleukin-2 receptor alpha chain; IL-2RA; IL-2Rα; Il2ra; IL-2R p55
Host Species Rat
Immunogen IL-2-dependent cytolytic mouse T-cell clone B6.1
Purification Method Affinity Chromatography
Quantity 0.1 mg
Regulatory Status RUO
Gene ID (Entrez) 16184
Target Species Mouse
Content And Storage Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze.
Form Liquid
Isotype IgG1 λ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.