Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBAK Monoclonal Antibody (6F9)
GREENER_CHOICE

Catalog No. PIMA522411
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA522411 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA522411 Supplier Invitrogen™ Supplier No. MA522411
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Peptide Sequence: TTKPNVIIKL EQGEEPWIMG GEFPCQHSPE AWRVDDLIER IQENEDKHSR QAACINSKTL TEEKENTFSQ IYMETSLVPS SIIAHNCVSC GKNLESISQL.

RBAK encodes a nuclear protein which interacts with the tumor suppressor retinoblastoma 1. The two interacting proteins are thought to act as a transcriptional repressor for promoters which are activated by the E2F1 transcription factor. This protein contains a Kruppel-associated box (KRAB), which is a transcriptional repressor motif.This gene encodes a nuclear protein which interacts with the tumor suppressor retinoblastoma 1. The two interacting proteins are thought to act as a transcriptional repressor for promoters which are activated by the E2F1 transcription factor. This protein contains a Kruppel-associated box (KRAB), which is a transcriptional repressor motif.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBAK
Applications ELISA, Western Blot
Classification Monoclonal
Clone 6F9
Concentration 0.2-1.0 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene RBAK
Gene Accession No. Q9NYW8
Gene Alias hRBaK; RB associated KRAB zinc finger; RBAK; RB-associated KRAB repressor; RB-associated KRAB zinc finger; RB-associated KRAB zinc finger protein; RGD1562875; Zinc finger protein 769; ZNF769
Gene Symbols RBAK
Host Species Mouse
Immunogen RBAK (NP_066986, 51 a.a. ~ 150 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 57786
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG2a κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.