Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBAK Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA567159
Description
Immunogen sequence: EYNECMEALDN EAVFIAHKRA YIGEKPYEWN DSGPDFIQMS NFNAYQRSQM EMKPF Highest antigen sequence identity to the following orthologs - mouse 71%, rat 71%.
RBAK encodes a nuclear protein which interacts with the tumor suppressor retinoblastoma 1. The two interacting proteins are thought to act as a transcriptional repressor for promoters which are activated by the E2F1 transcription factor. This protein contains a Kruppel-associated box (KRAB), which is a transcriptional repressor motif.This gene encodes a nuclear protein which interacts with the tumor suppressor retinoblastoma 1. The two interacting proteins are thought to act as a transcriptional repressor for promoters which are activated by the E2F1 transcription factor. This protein contains a Kruppel-associated box (KRAB), which is a transcriptional repressor motif.
Specifications
RBAK | |
Polyclonal | |
Unconjugated | |
RBAK | |
hRBaK; RB associated KRAB zinc finger; RBAK; RB-associated KRAB repressor; RB-associated KRAB zinc finger; RB-associated KRAB zinc finger protein; RGD1562875; Zinc finger protein 769; ZNF769 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
57786 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Immunocytochemistry | |
0.4 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9NYW8 | |
RBAK | |
Recombinant Human RBAK. Recombinant protein control fragment (Product #RP-107807). | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction