Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RbAp46 Antibody (OTI5A4), DyLight 755, Novus Biologicals™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals NBP273822IR
Description
RbAp46 Monoclonal specifically detects RbAp46 in Human, Mouse, Rat samples. It is validated for Western Blot, Immunohistochemistry.Specifications
RbAp46 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
RBBP7 | |
Full length recombinant protein of human RbAp46 (NP_002884) produced in HEK293T cells. | |
0.1 mL | |
Primary | |
Human, Mouse, Rat | |
IgG1 |
Western Blot, Immunohistochemistry | |
OTI5A4 | |
Western Blot, Immunohistochemistry | |
G1/S transition control protein-binding protein RbAp46, Histone acetyltransferase type B subunit 2, histone-binding protein RBBP7, MGC138867, Nucleosome-remodeling factor subunit RBAP46, RbAp46, RBBP-7, retinoblastoma binding protein 7, Retinoblastoma-binding protein 7MGC138868, Retinoblastoma-binding protein p46, retinoblastoma-binding protein RbAp46 | |
Mouse | |
Affinity Purified | |
Cancer, Chromatin Research, Growth and Development, Stem Cell Markers, Tumor Suppressors | |
5931.0 | |
Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction