Learn More
Invitrogen™ RbAp46 Monoclonal Antibody (OTI3A12)

Description
RbAp46 Monoclonal Antibody for Western Blot, ICC/IF
Specifications
Specifications
| Antigen | RbAp46 |
| Applications | Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI3A12 |
| Concentration | 0.85 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RBBP7 |
| Gene Accession No. | Q16576 |
| Gene Alias | AA409861; AI173248; AU019541; BB114024; G1/S transition control protein-binding protein; G1/S transition control protein-binding protein RbAp46; histone acetyltransferase type B subunit 2; Histone-binding protein RBBP7; mRbAp46; Nucleosome-remodeling factor subunit RBAP46; RB binding protein 7, chromatin remodeling factor; Rbap46; Rbbp7; RBBP-7; retinoblastoma binding protein 7; retinoblastoma-binding protein 7; Retinoblastoma-binding protein p46; retinoblastoma-binding protein RbAp46 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.