Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RbAp46/RbAp48 Monoclonal Antibody (15G12)
GREENER_CHOICE

Catalog No. PIMA123277
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA123277 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA123277 Supplier Invitrogen™ Supplier No. MA123277
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Recommended positive controls: K562, THP-1, HL-60. Predicted molecular weight is 47.6kDa. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RbAp48-15G12 recognizes full length RbAp48 (48 kDa) and RbAp46 (46 kDa), related nuclear proteins first isolated by their ability to interact with the carboxyl-terminus of the retinoblastoma protein (Rb). These two proteins are highly homologous at the amino acid level and are related functionally to members of the WD (Trp-Asp) repeat family of proteins. RbAp48 is one of the three subunits of CAF-1, can bind to histone H4 and is a subunit of human histone deacetylase HD1. Both RbAp48 and RbAp46 interact with HDAC1 and mSin3 in vivo.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RbAp46/RbAp48
Applications Gel Shift, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 15G12
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 20% glycerol and no preservative; pH 7
Gene RBBP7
Gene Accession No. Q09028, Q16576, Q60972, Q60973
Gene Alias AA409861; AI173248; AU019541; BB114024; CAF-1 p48 subunit; CAF-1 subunit C; CAF-I 48 kDa subunit; CAF-I p48; chromatin assembly factor 1 subunit C; Chromatin assembly factor I p48 subunit; chromatin assembly factor/CAF-1 p48 subunit; G1/S transition control protein-binding protein; G1/S transition control protein-binding protein RbAp46; histone acetyltransferase type B subunit 2; Histone-binding protein RBBP4; histone-binding protein RBBP7; lin-53; mRbAp46; mRbAp48; MSI1 protein homolog; nucleosome-remodeling factor subunit RBAP46; Nucleosome-remodeling factor subunit RBAP48; NURF55; RB binding protein 4, chromatin remodeling factor; RB binding protein 7, chromatin remodeling factor; Rbap46; RBAP48; Rbbp4; RBBP-4; RBBP7; RBBP-7; retinoblastoma binding protein 4; retinoblastoma binding protein 7; retinoblastoma-binding protein 4; Retinoblastoma-binding protein 7; retinoblastoma-binding protein p46; Retinoblastoma-binding protein p48; retinoblastoma-binding protein RbAp46
Gene Symbols RBBP4, RBBP7
Host Species Mouse
Immunogen The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19646, 245688, 5928, 5931
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.