Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RbAp48 Monoclonal Antibody (11G10)
GREENER_CHOICE

Catalog No. MA123273
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA123273 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA123273 Supplier Invitrogen™ Supplier No. MA123273
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Positive control: MOLT4, HeLa, LB22. For ICC a permeabilization step is recommended; use at a dilution of 1:500-1:1000 in PBST containing 2%BSA and 10% normal serum at 4°C overnight. Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

RbAp48-11G10 recognizes full length RbAp48, a 48 kDa nuclear protein first isolated by its ability to interact with the carboxyl-terminus of the retinoblastoma protein (Rb). It is a member of the WD (Trp-Asp) repeat family of proteins. RbAp48 is one of the three subunits of CAF-1, can bind to histone H4 and is a subunit of human histone deacetylase HD1.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RbAp48
Applications ChIP Assay, Functional Assay, Immunohistochemistry (Paraffin), Immunoprecipitation, Neutralization, Western Blot, Immunocytochemistry
Classification Monoclonal
Clone 11G10
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative
Gene RBBP4
Gene Accession No. Q09028, Q60972
Gene Alias CAF-1 p48 subunit; CAF-1 subunit C; CAF-I 48 kDa subunit; CAF-I p48; chromatin assembly factor 1 subunit C; Chromatin assembly factor I p48 subunit; chromatin assembly factor/CAF-1 p48 subunit; histone-binding protein RBBP4; lin-53; mRbAp48; MSI1 protein homolog; nucleosome-remodeling factor subunit RBAP48; NURF55; RB binding protein 4, chromatin remodeling factor; RBAP48; RBBP4; RBBP-4; retinoblastoma binding protein 4; retinoblastoma-binding protein 4; Retinoblastoma-binding protein p48
Gene Symbols RBBP4
Host Species Mouse
Immunogen The complete coding region (amino acids 1-425) of RbAp48 expressed in E. coli.
Purification Method Protein G
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19646, 313048, 5928
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.