Learn More
Invitrogen™ RbAp48 Polyclonal Antibody

Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA530422
Description
Recommended positive controls: 293T, A431, Jurkat, SH-SY5Y, SH-SY5Y nuclear extract. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (97%), Xenopus laevis (98%), Chicken (100%), Rhesus Monkey (100%), Chimpanzee (100%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.
RbAp48-11G10 recognizes full length RbAp48, a 48 kDa nuclear protein first isolated by its ability to interact with the carboxyl-terminus of the retinoblastoma protein (Rb). It is a member of the WD (Trp-Asp) repeat family of proteins. RbAp48 is one of the three subunits of CAF-1, can bind to histone H4 and is a subunit of human histone deacetylase HD1.
Specifications
| RbAp48 | |
| Polyclonal | |
| Unconjugated | |
| RBBP4 | |
| CAF-1 p48 subunit; CAF-1 subunit C; CAF-I 48 kDa subunit; CAF-I p48; chromatin assembly factor 1 subunit C; Chromatin assembly factor I p48 subunit; chromatin assembly factor/CAF-1 p48 subunit; histone-binding protein RBBP4; lin-53; mRbAp48; MSI1 protein homolog; nucleosome-remodeling factor subunit RBAP48; NURF55; RB binding protein 4, chromatin remodeling factor; RBAP48; RBBP4; RBBP-4; retinoblastoma binding protein 4; retinoblastoma-binding protein 4; Retinoblastoma-binding protein p48 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 5928 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.28 mg/mL | |
| 0.1M tris glycine with 20% glycerol and 0.01% thimerosal; pH 7 | |
| Q09028 | |
| RBBP4 | |
| Recombinant protein encompassing a sequence within the center region of human RbAp48. The exact sequence is proprietary. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.