Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBBP 4/7 Antibody (968721), DyLight 755, R&D Systems™

Mouse Monoclonal Antibody
Supplier: Novus Biologicals MAB9457IR
Description
RBBP 4/7 Monoclonal specifically detects RBBP 4/7 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RBBP 4/7 | |
Monoclonal | |
DyLight 755 | |
50mM Sodium Borate with 0.05% Sodium Azide | |
E. coli-derived recombinant human RBBP-4 | |
100 μL | |
Primary | |
Store at 4C in the dark. | |
IgG1 |
Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
968721 | |
Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
Mouse | |
Protein A or G purified | |
RUO | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only (RUO)
Spot an opportunity for improvement?Share a Content Correction