Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBBP5 Polyclonal Antibody
Rabbit Polyclonal Antibody
Supplier: Invitrogen™ PA5101839
Description
Antibody detects endogenous levels of total RBBP5.
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. The encoded protein binds directly to retinoblastoma protein, which regulates cell proliferation. It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
Specifications
RBBP5 | |
Polyclonal | |
Unconjugated | |
Rbbp5 | |
4933411J24Rik; C330016J05; RB binding protein 5, histone lysine methyltransferase complex subunit; RBBP 5; Rbbp5; RBBP-5; RBQ 3; RBQ3; retinoblastoma binding protein 5; retinoblastoma-binding protein 5; Retinoblastoma-binding protein RBQ-3; SWD1; SWD1, Set1c WD40 repeat protein, homolog | |
Rabbit | |
Affinity Chromatography | |
RUO | |
213464, 304794, 5929 | |
-20°C | |
Liquid |
Western Blot, Immunocytochemistry | |
1 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
Q15291, Q8BX09 | |
Rbbp5 | |
A synthesized peptide derived from human RBBP5(Accession Q15291), corresponding to amino acid residues P420-L470. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Safety and Handling
WARNING: Cancer - www.P65Warnings.ca.gov
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction