Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBBP5 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA540843
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA540843 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA540843 Supplier Invitrogen™ Supplier No. PA540843
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Peptide sequence: GASSEKKRQS SADGSQPPKK KPKTTNIELQ GVPNDEVHPL LGVKGDGKSK Sequence homology: Cow: 100%; Dog: 100%; Horse: 92%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.

This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. The encoded protein binds directly to retinoblastoma protein, which regulates cell proliferation. It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBBP5
Applications Western Blot
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 2% sucrose and 0.09% sodium azide
Gene Rbbp5
Gene Accession No. Q15291
Gene Alias 4933411J24Rik; C330016J05; RB binding protein 5, histone lysine methyltransferase complex subunit; RBBP 5; Rbbp5; RBBP-5; RBQ 3; RBQ3; retinoblastoma binding protein 5; retinoblastoma-binding protein 5; Retinoblastoma-binding protein RBQ-3; SWD1; SWD1, Set1c WD40 repeat protein, homolog
Gene Symbols Rbbp5
Host Species Rabbit
Immunogen Synthetic peptide directed towards the middle region of human RBBP5.
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5929
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.