Learn More
RBBP5 Polyclonal Antibody, Active Motif™

Description
Specifications
Specifications
| Antigen | RBBP5 |
| Applications | ChIP Assay, ChIP sequencing (ChIP-seq), Western Blot |
| Classification | Polyclonal |
| Concentration | Lot-Specific |
| Conjugate | Unconjugated |
| Formulation | PBS with 30% glycerol and 0.035% sodium azide; pH 7.5 |
| Gene | Rbbp5 |
| Gene Accession No. | Q15291 |
| Gene Alias | 4933411J24Rik; BOS_15527; C330016J05; RB binding protein 5, histone lysine methyltransferase complex subunit; RBBP 5; Rbbp5; RBBP-5; RBQ 3; RBQ3; retinoblastoma binding protein 5; Retinoblastoma-binding protein 5; Retinoblastoma-binding protein RBQ-3; SWD1; SWD1, Set1c WD40 repeat protein, homolog |
| Gene Symbols | Rbbp5 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.