Learn More
RbBP5 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
$151.31 - $568.26
Specifications
Antigen | RbBP5 |
---|---|
Applications | Immunohistochemistry |
Classification | Polyclonal |
Conjugate | Unconjugated |
Form | Liquid |
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Quantity | Price | Quantity & Availability | |||||
50-157-1618
![]() |
Bethyl Laboratories, Inc
IHC00172T |
10 μL |
Each for $151.31
|
|
|||||
50-157-1617
![]() |
Bethyl Laboratories, Inc
IHC00172 |
100 μL |
Each for $568.26
|
|
|||||
Description
The recommended shelf life for this product is 1 year from date of receipt. Based on 100% sequence identity, this antibody is predicted to react with Bovine, Horse, Orangutan, Monkey, Gorilla, Northern white-cheeked gibbon, Little brown bat and hi
The protein encoded by this gene is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that bind directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B.Specifications
RbBP5 | |
Polyclonal | |
Liquid | |
RUO | |
TBS with 0.1% BSA and 0.09% sodium azide | |
RBQ3, SWD1, RBBP-5, SWD1, Set1c WD40 repeat protein, homolog, retinoblastoma-binding protein 5, retinoblastoma-binding protein RBQ-3 | |
Between 500 and C-term | |
Primary | |
4° C | |
RBBP5 |
Immunohistochemistry | |
Unconjugated | |
Rabbit | |
Human, Mouse | |
Q15291, Q8BX09 | |
213464, 5929 | |
IgG | |
Antigen affinity chromatography | |
Antibody |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.