Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBBP5 Recombinant Rabbit Monoclonal Antibody (PSH0-15)
GREENER_CHOICE

Catalog No. FEMA550779
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEMA550779 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEMA550779 Supplier Invitrogen™ Supplier No. MA550779
Add to Cart
Add to Cart

Rabbit Recombinant Monoclonal Antibody

Positive control: SW480 cell lysate, A431 cell lysate, C2C12 cell lysate, 293 cell lysate, MCF-7 cell lysate, NIH/3T3 cell lysate, K562 cell lysate, N2A cell lysate, PC-12 cell lysate, RAW264.7 cell lysate. Predicted band size: 59 kDa Subcellular Location: Nucleus.

This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. The encoded protein binds directly to retinoblastoma protein, which regulates cell proliferation. It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBBP5
Applications Western Blot
Classification Recombinant Monoclonal
Clone PSH0-15
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene Rbbp5
Gene Accession No. Q15291, Q8BX09
Gene Alias 4933411J24Rik; C330016J05; RB binding protein 5, histone lysine methyltransferase complex subunit; RBBP 5; Rbbp5; RBBP-5; RBQ 3; RBQ3; retinoblastoma binding protein 5; retinoblastoma-binding protein 5; Retinoblastoma-binding protein RBQ-3; SWD1; SWD1, Set1c WD40 repeat protein, homolog
Gene Symbols Rbbp5
Host Species Rabbit
Immunogen Synthetic peptide within human RbBP5 aa 431-480/538.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 213464, 304794, 5929
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.