Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBBP5 Recombinant Rabbit Monoclonal Antibody (PSH0-16)
Rabbit Recombinant Monoclonal Antibody
Supplier: Invitrogen™ MA550780
Description
Positive control: Human colon tissue, human colon carcinoma tissue, human mouse tissue. Predicted band size: 59 kDa Subcellular Location: Nucleus.
This gene encodes a ubiquitously expressed nuclear protein which belongs to a highly conserved subfamily of WD-repeat proteins. The encoded protein binds directly to retinoblastoma protein, which regulates cell proliferation. It interacts preferentially with the underphosphorylated retinoblastoma protein via the E1A-binding pocket B. Three alternatively spliced transcript variants that encode different protein isoforms have been described for this gene.
Specifications
RBBP5 | |
Recombinant Monoclonal | |
1 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
Q15291, Q8BX09 | |
Rbbp5 | |
Synthetic peptide within human RbBP5 aa 431-480/538. | |
100 μL | |
Primary | |
Human, Mouse | |
Antibody | |
IgG |
Immunohistochemistry (Paraffin) | |
PSH0-16 | |
Unconjugated | |
Rbbp5 | |
4933411J24Rik; C330016J05; RB binding protein 5, histone lysine methyltransferase complex subunit; RBBP 5; Rbbp5; RBBP-5; RBQ 3; RBQ3; retinoblastoma binding protein 5; retinoblastoma-binding protein 5; Retinoblastoma-binding protein RBQ-3; SWD1; SWD1, Set1c WD40 repeat protein, homolog | |
Rabbit | |
Protein A | |
RUO | |
213464, 5929 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction