Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBBP6 Antibody, mFluor Violet 500 SE, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP149542MFV500
Description
RBBP6 Polyclonal antibody specifically detects RBBP6 in Human samples. It is validated for Western Blot, ImmunoprecipitationSpecifications
RBBP6 | |
Polyclonal | |
Western Blot, Immunoprecipitation | |
E3 ubiquitin-protein ligase RBBP6, EC 6.3.2.-, MY038, P2PR, P2P-R, p53-associated cellular protein of testis, PACTDKFZp761B2423, Proliferation potential-related protein, Protein P2P-R, RB-binding Q-protein 1, RBQ1, RBQ-1, retinoblastoma binding protein 6, Retinoblastoma-binding protein 6DKFZp686P0638, Retinoblastoma-binding Q protein 1, SNAMA | |
A synthetic peptide made to an internal portion of the human RBBP6 protein (between residues 20-70) [UniProt Q7Z6E9] | |
0.1 mL | |
Tumor Suppressors | |
5930 | |
Store at 4C in the dark. | |
IgG |
Western Blot, Immunoprecipitation | |
mFluor Violet 500 SE | |
50mM Sodium Borate | |
Rabbit | |
Immunogen affinity purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction