Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBBP6 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA5104100
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA5104100 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA5104100 Supplier Invitrogen™ Supplier No. PA5104100
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Antibody detects endogenous levels of total RBBP6.

This gene encodes a cilia-associated protein belonging to the kinesin family. This protein plays a role in the sonic hedgehog (SHH) signaling pathway through the regulation of GLI transcription factors. It functions as a negative regulator of the SHH pathway by preventing inappropriate activation of GLI2 in the absence of ligand, and as a positive regulator by preventing the processing of GLI3 into its repressor form. Mutations in this gene have been associated with various ciliopathies.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBBP6
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
Gene Rbbp6
Gene Accession No. P97868, Q7Z6E9
Gene Alias 4933422O15Rik; AI316869; BB233631; C030034J04Rik; C77662; E3 ubiquitin-protein ligase RBBP6; im:7158869; MY038; my038-like; P2PR; P2P-R; p53-associated cellular protein of testis; PACT; Proliferation potential-related protein; protein P2P-R; RB binding protein 6, ubiquitin ligase; RB-binding Q-protein 1; Rbbp6; rbbp6l; RBQ1; RBQ-1; retinoblastoma binding protein 6; retinoblastoma-binding protein 6; retinoblastoma-binding protein 6, isoform 3; retinoblastoma-binding Q protein 1; RING-type E3 ubiquitin transferase RBBP6; SNAMA
Gene Symbols Rbbp6
Host Species Rabbit
Immunogen A synthesized peptide derived from human RBBP6(Accession Q7Z6E9), corresponding to amino acid residues F1440-E1490.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 19647, 5930
Target Species Human, Mouse
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.