Learn More
RBBP6 Polyclonal Antibody, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304975AT

Description
The recommended shelf life for this product is 1 year from date of receipt.
The retinoblastoma tumor suppressor (pRB) protein binds with many other proteins. In various human cancers, pRB suppresses cellular proliferation and is inactivated. Cell cycle-dependent phosphorylation regulates the activity of pRB. This gene encodes a protein which binds to underphosphorylated but not phosphorylated pRB. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene.Specifications
RBBP6 | |
Polyclonal | |
TBS with 0.1% BSA and 0.09% sodium azide | |
Q7Z6E9 | |
RBBP6 | |
Between 50 and 100 | |
20 μL | |
Primary | |
Human | |
Antibody | |
IgG |
Western Blot | |
Unconjugated | |
RBBP6 | |
My038, MY038, P2P-R, PACT, RBQ-1, SNAMA, E3 ubiquitin-protein ligase RBBP6, RB-binding Q-protein 1, p53-associated cellular protein of testis, proliferation potential-related protein, protein P2P-R, retinoblastoma-binding Q protein 1, retinoblastoma-binding protein 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5930 | |
4° C | |
Liquid |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.