Learn More
RBBP6 Rabbit anti-Human, Polyclonal, Bethyl Laboratories
Rabbit Polyclonal Antibody
Supplier: Bethyl Laboratories, Inc A304976A

Description
The recommended shelf life for this product is 1 year from date of receipt.
The retinoblastoma tumor suppressor (pRB) protein binds with many other proteins. In various human cancers, pRB suppresses cellular proliferation and is inactivated. Cell cycle-dependent phosphorylation regulates the activity of pRB. This gene encodes a protein which binds to underphosphorylated but not phosphorylated pRB. Multiple alternatively spliced transcript variants that encode different isoforms have been found for this gene.Specifications
RBBP6 | |
Polyclonal | |
Unconjugated | |
RBBP6 | |
My038, MY038, P2P-R, PACT, RBQ-1, SNAMA, E3 ubiquitin-protein ligase RBBP6, RB-binding Q-protein 1, p53-associated cellular protein of testis, proliferation potential-related protein, protein P2P-R, retinoblastoma-binding Q protein 1, retinoblastoma-binding protein 6 | |
Rabbit | |
Antigen affinity chromatography | |
RUO | |
5930 | |
4° C | |
Liquid |
Immunoprecipitation | |
1 mg/ml | |
phosphate, tris citrate with 0.09% sodium azide; pH 7-8 | |
Q7Z6E9 | |
RBBP6 | |
Between 900 and 950 | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.