Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP7 Monoclonal Antibody (OTI3A12), TrueMAB™, OriGene
SDP

Catalog No. 502540696
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-0696 30 μL
50-167-4610 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-0696 Supplier OriGene Supplier No. TA503809S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBBP7 Monoclonal antibody specifically detects RBBP7 in Human samples. It is validated for Immunocytochemistry, Western Blot

Specifications

Antigen RBBP7
Applications Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI3A12
Concentration 0.85 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene Katnb1
Gene Accession No. Q9BVA0
Gene Alias AA409861; AI173248; AU019541; BB114024; G1/S transition control protein-binding protein; G1/S transition control protein-binding protein RbAp46; histone acetyltransferase type B subunit 2; Histone-binding protein RBBP7; mRbAp46; Nucleosome-remodeling factor subunit RBAP46; RB binding protein 7, chromatin remodeling factor; Rbap46; Rbbp7; RBBP-7; retinoblastoma binding protein 7; retinoblastoma-binding protein 7; Retinoblastoma-binding protein p46; retinoblastoma-binding protein RbAp46
Host Species Mouse
Immunogen Full length human recombinant protein of human KATNB1 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10300
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.