Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP7 Mouse anti-Human, Clone: OTI2D8, liquid, TrueMAB™
SDP

Catalog No. 501677455
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-167-7455 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-167-7455 Supplier OriGene Supplier No. TA600416
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

This protein is a ubiquitously expressed nuclear protein and belongs to a highly conserved subfamily of WD-repeat proteins. It is found among several proteins that binds directly to retinoblastoma protein, which regulates cell proliferation. The encoded protein is found in many histone deacetylase complexes, including mSin3 co-repressor complex. It is also present in protein complexes involved in chromatin assembly. This protein can interact with BRCA1 tumor-suppressor gene and may have a role in the regulation of cell proliferation and differentiation.

Specifications

Antigen RBBP7
Applications ELISA
Classification Monoclonal
Clone OTI2D8
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with <5% trehalose and 0.05% sodium azide
Gene RBBP7
Gene Accession No. Q16576
Gene Alias RbAp46
Gene Symbols RBBP7
Host Species Mouse
Immunogen Full length human recombit protein of human RBBP7 produced in HEK293T cell.
Purification Method Affinity Chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 5931
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.