Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP7 Polyclonal Antibody, Huabio

Catalog No. 502544598
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-4598 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-254-4598 Supplier HUABIO RESEARCH INC Supplier No. HA500155
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

RBBP7 Polyclonal antibody specifically detects RBBP7 in Human, Mouse samples. It is validated for Immunohistochemistry (Paraffin), Western Blot

Specifications

Antigen RBBP7
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation TBS with 0.2% BSA, 50% glycerol and 0.05% sodium azide; pH 7.4
Gene RBBP7
Gene Accession No. Q16576, Q60973
Gene Alias AA409861; AI173248; AU019541; BB114024; G1/S transition control protein-binding protein; G1/S transition control protein-binding protein RbAp46; histone acetyltransferase type B subunit 2; Histone-binding protein RBBP7; mRbAp46; Nucleosome-remodeling factor subunit RBAP46; RB binding protein 7, chromatin remodeling factor; Rbap46; Rbbp7; RBBP-7; retinoblastoma binding protein 7; retinoblastoma-binding protein 7; Retinoblastoma-binding protein p46; retinoblastoma-binding protein RbAp46
Host Species Rabbit
Immunogen Recombinant protein within human RBBP7 aa 225-425.
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 245688, 5931
Target Species Human, Mouse
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.