Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

RBBP9 Monoclonal Antibody (OTI1C10), TrueMAB™, OriGene
SDP

Catalog No. 502542104
Encompass
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μL
30 μL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-254-2104 30 μL
50-167-2801 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 50-254-2104 Supplier OriGene Supplier No. TA501677S
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBBP9 Monoclonal antibody specifically detects RBBP9 in Human samples. It is validated for Flow Cytometry, Immunocytochemistry, Western Blot

Specifications

Antigen RBBP9
Applications Flow Cytometry, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone OTI1C10
Concentration 0.29 mg/mL
Conjugate Unconjugated
Formulation PBS with 1% BSA, 50% glycerol and 0.02% sodium azide
Gene RBBP9
Gene Accession No. O75884
Gene Alias B5T overexpressed gene protein; B5T-overexpressed gene protein; BOG; im:7247963; im:7247963 protein; MGC9236; Protein BOG; Putative hydrolase RBBP9; RB binding protein 9, serine hydrolase; RBBP10; RBBP-10; Rbbp9; RBBP-9; retinoblastoma binding protein 9; retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; serine hydrolase RBBP9; serine hydrolase RBBP9; putative hydrolase RBBP9; si:ch211-258l4.7; zgc:152807
Host Species Mouse
Immunogen Full length human recombinant protein of human RBBP9 produced in HEK293T cell.
Purification Method Affinity chromatography
Quantity 30 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10741
Target Species Human
Content And Storage -20° C, Avoid Freeze/Thaw Cycles
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.