Learn More
Invitrogen™ RBBP9 Monoclonal Antibody (OTI4G9)

Description
RBBP9 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P), Flow
Specifications
Specifications
| Antigen | RBBP9 |
| Applications | Flow Cytometry, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Monoclonal |
| Clone | OTI4G9 |
| Concentration | 0.67 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 |
| Gene | RBBP9 |
| Gene Accession No. | O75884 |
| Gene Alias | B5T overexpressed gene protein; B5T-overexpressed gene protein; BOG; im:7247963; im:7247963 protein; MGC9236; Protein BOG; Putative hydrolase RBBP9; RB binding protein 9, serine hydrolase; RBBP10; RBBP-10; Rbbp9; RBBP-9; retinoblastoma binding protein 9; retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; serine hydrolase RBBP9; serine hydrolase RBBP9; putative hydrolase RBBP9; si:ch211-258l4.7; zgc:152807 |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.