Learn More
RBBP9 Mouse anti-Human, Clone: OTI4E1, lyophilized, TrueMAB™

Mouse Monoclonal Antibody
Supplier: OriGene CF501740

Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.Specifications
RBBP9 | |
Monoclonal | |
Unconjugated | |
RBBP9 | |
BOG, RBBP10 | |
Mouse | |
Affinity Chromatography | |
RUO | |
10741 | |
-20° C, Avoid Freeze/Thaw Cycles | |
Lyophilized |
Immunocytochemistry, Immunofluorescence, Western Blot | |
OTI4E1 | |
PBS with 8% trehalose and no preservative; pH 7.3 | |
O75884 | |
RBBP9 | |
Full length human recombit protein of human RBBP9 produced in HEK293T cell. | |
100 μg | |
Primary | |
Human | |
Antibody | |
IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.