Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBBP9 Polyclonal Antibody
GREENER_CHOICE

Catalog No. PIPA553389
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA553389 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA553389 Supplier Invitrogen™ Supplier No. PA553389
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Immunogen sequence: MASPSKAVIV PGNGGGDVTT HGWYGWVKKE LEKIPGFQCL AKNMPDPITA RESIWLPFME TELHCDEKTI IIGHSSGAIA AMRYAETHRV YAIVLVSAYT SDLGDENERA S Highest antigen sequence identity to the following orthologs: Mouse - 95%, Rat - 93%.

RBBP9 may play a role in the transformation process due to its capacity to confer resistance to the growth-inhibitory effects of TGF-beta1 through interaction with retinoblastoma and the subsequent displacement of E2F-1.The protein encoded by this gene is a retinoblastoma binding protein that may play a role in the regulation of cell proliferation and differentiation. Two alternatively spliced transcript variants of this gene with identical predicted protein products have been reported, one of which is a nonsense-mediated decay candidate.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBBP9
Applications Immunohistochemistry (Paraffin), Western Blot
Classification Polyclonal
Concentration 0.05 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene RBBP9
Gene Accession No. O75884
Gene Alias B5T overexpressed gene protein; B5T-overexpressed gene protein; BOG; im:7247963; im:7247963 protein; MGC9236; Protein BOG; Putative hydrolase RBBP9; RB binding protein 9, serine hydrolase; RBBP10; RBBP-10; Rbbp9; RBBP-9; retinoblastoma binding protein 9; retinoblastoma-binding protein 10; Retinoblastoma-binding protein 9; retinoma-binding protein 9; serine hydrolase RBBP9; serine hydrolase RBBP9; putative hydrolase RBBP9; si:ch211-258l4.7; zgc:152807
Gene Symbols RBBP9
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human RBBP9. Recombinant protein control fragment (Product #RP-91606).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10741
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.