Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBC Monoclonal Antibody (34-3C)
GREENER_CHOICE

Catalog No. FERMA549100
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FERMA549100 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FERMA549100 Supplier Invitrogen™ Supplier No. MA549100
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

The monoclonal antibody only recognises antigenic determinants expressed on Mouse RBC and not on other species of RBC.

Red blood cells are the most common type of blood cell and are the vertebrate body's principal means of delivering oxygen from the lungs or gills to body tissues via the blood.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBC
Applications Flow Cytometry, Functional Assay
Classification Monoclonal
Clone 34-3C
Concentration 100 μg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA and no preservative
Gene Alias Red Blood Cell
Host Species Mouse
Immunogen RBC.
Purification Method Protein G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Target Species Mouse
Content And Storage 4°C
Product Type Antibody
Form Liquid
Isotype IgG2a
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.