Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
RBCK1 Antibody, Alexa Fluor™ 532, Novus Biologicals™

Rabbit Polyclonal Antibody
Supplier: Novus Biologicals NBP298085AF532
Description
RBCK1 Polyclonal specifically detects RBCK1 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.Specifications
RBCK1 | |
Polyclonal | |
Immunohistochemistry, Immunohistochemistry-Paraffin | |
C20orf18, EC 6.3.2.-, HBV associated factor 4, HBV-associated factor 4, Heme-oxidized IRP2 ubiquitin ligase 1, Hepatitis B virus X-associated protein 4, HOIL-1, RanBP-type and C3HC4-type zinc finger containing 1, ranBP-type and C3HC4-type zinc finger-containing protein 1, RBCC protein interacting with PKC1, RBCK2, RNF54RING finger protein 54, UBCE7IP3HOIL1, ubiquitin conjugating enzyme 7 interacting protein 3, Ubiquitin-conjugating enzyme 7-interacting protein 3, XAP3, XAP4chromosome 20 open reading frame 18, ZRANB4 | |
Produced in rabbits immunized with E. coli-derived Human RBCK1 fragment. | |
0.1 mL | |
Zinc Finger | |
10616 | |
Store at 4°C in the dark. | |
IgG |
Immunohistochemistry, Immunohistochemistry (Paraffin) | |
Alexa Fluor 532 | |
50mM Sodium Borate | |
Rabbit | |
Antigen Affinity-purified | |
RUO | |
Primary | |
Human | |
Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction