Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ RBCK1 Monoclonal Antibody (1839CT718.19.54)
GREENER_CHOICE

Catalog No. PIMA537756
Change view
Click to view available options
Quantity:
200 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA537756 200 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA537756 Supplier Invitrogen™ Supplier No. MA537756
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

RBCK1 Monoclonal Antibody for Western Blot

RBCK1 encodes a protein that is similar to mouse UIP28/UbcM4 interacting protein. Alternative splicing has been observed at this locus, resulting in distinct isoforms.
TRUSTED_SUSTAINABILITY

Specifications

Antigen RBCK1
Applications Western Blot
Classification Monoclonal
Clone 1839CT718.19.54
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.4
Gene RBCK1
Gene Accession No. Q9BYM8
Gene Alias AL033326; C20orf18; HBV-associated factor 4; heme-oxidized IRP2 ubiquitin ligase 1; Heme-oxidized IRP2 ubiquitin ligase 1 homolog; hepatitis B virus X-associated protein 4; HOIL1; HOIL-1; HOIL-1L; novel zinc finger, C3HC4 type (RING finger) protein; PBMEI; PGBM1; Pkcbpb15; protein kinase C-binding protein Beta15; protein kinase C-binding protein beta-15; RANBP2-type and C3HC4-type zinc finger containing 1; RanBP-type and C3HC4-type zinc finger containing 1; ranBP-type and C3HC4-type zinc finger-containing protein 1; RBCC protein interacting with PKC; RBCC protein interacting with PKC 1; RBCC protein interacting with PKC1; Rbck; RBCK1; RBCK2; RING finger protein 54; RING-type E3 ubiquitin transferase HOIL-1; RNF54; UBCE7IP3; ubcM4-interacting protein 28; ubiquitin conjugating enzyme 7 interacting protein 3; ubiquitin-conjugating enzyme 7-interacting protein 3; Uip28; XAP3; XAP4; ZRANB4
Gene Symbols RBCK1
Host Species Mouse
Immunogen recombinant protein of human RBCK1 (UBCE7IP3).
Purification Method Protein G
Quantity 200 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 10616
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1 κ
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.