Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Invitrogen™ RBCK1 Monoclonal Antibody (CL4289)
Mouse Monoclonal Antibody
Supplier: Invitrogen™ MA531400
Description
Immunogen sequence: SVEDAQMHTV TIWLTVRPDM TVASLKDMVF LDYGFPPVLQ QWVIGQRLAR DQETLHSHGV RQNGDSAYLY LLSARNTSLN PQELQRERQL RMLEDLGFKD LTLQPRGPLE PGPPKPGVPQ EPGRGQPDAV PEP.
RBCK1 encodes a protein that is similar to mouse UIP28/UbcM4 interacting protein. Alternative splicing has been observed at this locus, resulting in distinct isoforms.
Specifications
RBCK1 | |
Monoclonal | |
1 mg/mL | |
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
Q9BYM8 | |
RBCK1 | |
Recombinant protein corresponding to Human RBCK1. | |
100 μL | |
Primary | |
Human | |
Antibody | |
IgG1 |
Immunohistochemistry (Paraffin), Western Blot | |
CL4289 | |
Unconjugated | |
RBCK1 | |
AL033326; C20orf18; HBV-associated factor 4; heme-oxidized IRP2 ubiquitin ligase 1; Heme-oxidized IRP2 ubiquitin ligase 1 homolog; hepatitis B virus X-associated protein 4; HOIL1; HOIL-1; HOIL-1L; novel zinc finger, C3HC4 type (RING finger) protein; PBMEI; PGBM1; Pkcbpb15; protein kinase C-binding protein Beta15; protein kinase C-binding protein beta-15; RANBP2-type and C3HC4-type zinc finger containing 1; RanBP-type and C3HC4-type zinc finger containing 1; ranBP-type and C3HC4-type zinc finger-containing protein 1; RBCC protein interacting with PKC; RBCC protein interacting with PKC 1; RBCC protein interacting with PKC1; Rbck; RBCK1; RBCK2; RING finger protein 54; RING-type E3 ubiquitin transferase HOIL-1; RNF54; UBCE7IP3; ubcM4-interacting protein 28; ubiquitin conjugating enzyme 7 interacting protein 3; ubiquitin-conjugating enzyme 7-interacting protein 3; Uip28; XAP3; XAP4; ZRANB4 | |
Mouse | |
Protein A | |
RUO | |
10616 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction